Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR7 Antibody (437310), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4405PECY7
Description
mGluR7 Monoclonal antibody specifically detects mGluR7 in Human samples. It is validated for Flow CytometrySpecifications
mGluR7 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
437310 | |
Flow Cytometry | |
FLJ40498, glutamate receptor, metabotropic 7, metabotropic glutamate receptor 7, mGlu7, mGluR7, MGLUR7GPRC1GGLUR7 | |
HEK293 human embryonic kidney cell line transfected with human mGluR7, Gln35-Ile915, Accession # Q14831 | |
0.1 mL | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
2917 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction