Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115575 Supplier Invitrogen™ Supplier No. PA5115575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total mGluR7.

L-glutamate is the major excitatory neurotransmitter in the central nervous system, and it activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into three groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5, and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3, while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GRM7
Gene Accession No. P35400, Q14831, Q68ED2
Gene Alias 6330570A01Rik; BB176677; C030018L03; E130018M02Rik; FLJ40498; G protein-coupled receptor GRM7; G protein-coupled receptor, family C, group 1, member G; GLUR7; glutamate metabotropic receptor 7; glutamate receptor; glutamate receptor, metabotropic 7; Gpr1g; Gprc1g; GRM7; Metabotropic glutamate receptor 7; metabotropic glutamate receptor mGluR7; metabotropic glutamate receptor subtype 7b; MGLU7; mGlu7a receptor; MGLUR7; PPP1R87; protein phosphatase 1, regulatory subunit 87
Gene Symbols GRM7
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRM7(Accession Q14831), corresponding to amino acid residues S874-I915.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108073, 2917, 81672
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.