Learn More
Invitrogen™ mGluR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120843
Description
Positive test controls include: K-562, 22Rv1, HepG2, HeLa. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: MVCEGKRSAS CPCFFLLTAK FYWILTMMQR THSQEYAHSI RVDGDIILGG LFPVHAKGER GVPCGELKKE KGIHRLEAML YAIDQINKDP DLLSNITLGV.
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities.
Specifications
| mGluR8 | |
| Polyclonal | |
| Unconjugated | |
| GRM8 | |
| A230002O04; FLJ41058; G protein coupled receptor, family C, group 1, member H; G protein-coupled receptor family C group 1 member H; G protein-coupled receptor, family C, group 1, member H; GLUR8; glutamate metabotropic receptor 8; glutamate receptor; glutamate receptor, metabotropic 8; Gprc1h; Grm8; M mGlu8; Metabotropic glutamate receptor 8; metabotropic glutamate receptor subtype 8b; MGC126724; mGlu8; mGlur; mGluR8; mGluR8b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2918 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.64 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00222 | |
| GRM8 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GRM8 (NP_0008362). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.