Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511697
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA511697 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA511697 Supplier Invitrogen™ Supplier No. PA511697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology.

L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR8
Applications Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GRM8
Gene Accession No. O00222, P47743
Gene Alias A230002O04; FLJ41058; G protein coupled receptor, family C, group 1, member H; G protein-coupled receptor family C group 1 member H; G protein-coupled receptor, family C, group 1, member H; GLUR8; glutamate metabotropic receptor 8; glutamate receptor; glutamate receptor, metabotropic 8; Gprc1h; Grm8; M mGlu8; Metabotropic glutamate receptor 8; metabotropic glutamate receptor subtype 8b; MGC126724; mGlu8; mGlur; mGluR8; mGluR8b
Gene Symbols GRM8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 852-882 amino acids from the C-terminal region of human Metabotropic Glutamate Receptor 8.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14823, 2918
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.