Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGME1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734011 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4011 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4011 Supplier Proteintech Group Inc Supplier No. 231781AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects the 35-40 kDa protein.

The function of this protein remains unknown.

Specifications

Antigen MGME1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MGME1
Gene Accession No. Q9BQP7, Q9CXC3
Gene Alias C20orf72, DDK1, MGME1
Gene Symbols Mgme1
Host Species Rabbit
Immunogen MGME1 Fusion Protein Ag19644
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74528, 92667
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.