Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599442 Supplier Invitrogen™ Supplier No. PA599442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MGMT.

O6-Methylguanine-DNA Methyltransferase (MGMT) is an important DNA repair protein involved in tumor cell resistance to the cytostatic activity of chemotherapeutic alkylating agents. This protein is also effective in protecting normal cells against the genotoxic and carcinogenic effects of DNA alkylation. The alkylating drug resistance is caused by MGMT's ability to remove DNA alkyl groups introduced in the O6 position of guanine. MGMT is expressed in highly variable amounts, depending upon the cell and tissue type, species, and cellular growth characteristics. In addition, MGMT activity varies among groups of tumors and within a particular type of tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGMT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MGMT
Gene Accession No. P16455, P24528, P26187
Gene Alias 0-6-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase; Agat; AGT; AI267024; methylated DNA protein cysteine methyltransferase; methylated-DNA--protein-cysteine methyltransferase; methylguanine-DNA methyltransferase; MGMT; O(6)-alkylguanine-DNA alkyltransferase; O-6-alkylguanine-DNA alkyltransferase; O6-alkylguanine-DNA alkyltransferase; O-6-methylguan; O6-methylguanine-DNA methyltranferase; O-6-methylguanine-DNA methyltransferase; O6-methylguanine-DNA methyltransferase; O-6-methylguanine-DNA-alkyltransferase
Gene Symbols MGMT
Host Species Rabbit
Immunogen A synthesized peptide derived from human MGMT(Accession P16455), corresponding to amino acid residues M1-K36.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17314, 25332, 4255
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.