Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119317
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119317 200 μL
PIPA5121775 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119317 Supplier Invitrogen™ Supplier No. PA5119317
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MGMT Polyclonal Antibody for Western Blot, IHC (P)

O6-Methylguanine-DNA Methyltransferase (MGMT) is an important DNA repair protein involved in tumor cell resistance to the cytostatic activity of chemotherapeutic alkylating agents. This protein is also effective in protecting normal cells against the genotoxic and carcinogenic effects of DNA alkylation. The alkylating drug resistance is caused by MGMT's ability to remove DNA alkyl groups introduced in the O6 position of guanine. MGMT is expressed in highly variable amounts, depending upon the cell and tissue type, species, and cellular growth characteristics. In addition, MGMT activity varies among groups of tumors and within a particular type of tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGMT
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MGMT
Gene Accession No. P16455
Gene Alias 0-6-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase; Agat; AGT; AI267024; methylated DNA protein cysteine methyltransferase; methylated-DNA--protein-cysteine methyltransferase; methylguanine-DNA methyltransferase; MGMT; O(6)-alkylguanine-DNA alkyltransferase; O-6-alkylguanine-DNA alkyltransferase; O6-alkylguanine-DNA alkyltransferase; O-6-methylguan; O6-methylguanine-DNA methyltranferase; O-6-methylguanine-DNA methyltransferase; O6-methylguanine-DNA methyltransferase; O-6-methylguanine-DNA-alkyltransferase
Gene Symbols MGMT
Host Species Rabbit
Immunogen Recombinant protein O-6-Methylguanine DNA Methyltransferase. The antigen corresponds to amino acid range 1-207 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.