Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGP Antibody (OTI11G6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272660H
Description
MGP Monoclonal specifically detects MGP in Human samples. It is validated for Western Blot.Specifications
MGP | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MGP | |
Full length recombinant protein of human MGP (NP_000891) produced in E.coli. | |
0.1 mL | |
4256.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI11G6 | |
Western Blot | |
Cell growth-inhibiting gene 36 protein, matrix Gla protein, MGLAP, NTI | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction