Learn More
Invitrogen™ MGP Monoclonal Antibody (OTI8D6)

Description
MGP Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MGP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI8D6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MGP |
| Gene Accession No. | P08493 |
| Gene Alias | cell growth-inhibiting gene 36 protein; GIG36; MATRIX; matrix gamma-carboxyglutamate (gla) protein; matrix Gla protein; Matrix Gla protein long form; Matrix Gla protein short form; Mglap; Mgp; MGP precursor; NTI; similar to matrix Gla protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.