Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGP Mouse anti-Human, Mouse, Clone: 1A1C3, Proteintech
SDP

Catalog No. 501736210 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6210 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6210 Supplier Proteintech Group Inc Supplier No. 600551IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.

Specifications

Antigen MGP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1A1C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MGP
Gene Accession No. P08493, P19788
Gene Alias GIG36, matrix Gla protein, MGLAP, MGP, NTI
Gene Symbols Mgp
Host Species Mouse
Immunogen MGP Fusion Protein Ag1091
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17313, 4256
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.