Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579669
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579669 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579669 Supplier Invitrogen™ Supplier No. PA579669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: JURKAT whole cell. IHC: human lung cancer tissue.

MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene MGP
Gene Accession No. P08493
Gene Alias cell growth-inhibiting gene 36 protein; GIG36; MATRIX; matrix gamma-carboxyglutamate (gla) protein; matrix Gla protein; Matrix Gla protein long form; Matrix Gla protein short form; Mglap; Mgp; MGP precursor; NTI; similar to matrix Gla protein
Gene Symbols MGP
Host Species Rabbit
Immunogen E.coli-derived human MGP recombinant protein (Position: Y20-F96).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4256
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.