Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MGP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596323
Description
Immunogen sequence: YESHESMESY ELNPFINRRN ANTFISPQQR WRAKVQERIR ERSKPVHELN REACDDYRLC ERYAMVYGYN AAYNRYFRKR RGTK; Positive Samples: MCF7, U-87MG, A375, BT-474, 3T3L1, Mouse kidney, Mouse lung, Mouse heart.
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.
Specifications
MGP | |
Polyclonal | |
Unconjugated | |
MGP | |
cell growth-inhibiting gene 36 protein; GIG36; MATRIX; matrix gamma-carboxyglutamate (gla) protein; matrix Gla protein; Matrix Gla protein long form; Matrix Gla protein short form; Mglap; Mgp; MGP precursor; NTI; similar to matrix Gla protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
17313, 25333, 4256 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.88 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P08493, P08494, P19788 | |
MGP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-103 of human MGP (NP_0008912). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction