Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGRN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217286
Description
MGRN1 Polyclonal specifically detects MGRN1 in Human samples. It is validated for Western Blot.Specifications
MGRN1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
E3 ubiquitin-protein ligase MGRN1, EC 6.3.2, KIAA0544probable E3 ubiquitin-protein ligase MGRN1, Mahogunin RING finger protein 1, mahogunin, ring finger 1, RING finger protein 156, RNF156EC 6.3.2.- | |
Rabbit | |
63 kDa | |
0.1 mL | |
Zinc Finger | |
23295 | |
Human | |
IgG |
Western Blot | |
0.71mg/mL | |
Western Blot 1:500-1:3000 | |
O60291 | |
MGRN1 | |
Recombinant protein encompassing a sequence within the center region of human MGRN1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (95%), Rat (95%), Xenopus laevis (81%), Zebrafish (81%), Bovine (95%), Xenopus tropicalis (81%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction