Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGRN1 Monoclonal Antibody (OTI1A11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502679S


Description
MGRN1 Monoclonal antibody specifically detects MGRN1 in Canine, Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
MGRN1 | |
Monoclonal | |
0.52 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O00338, Q9D939 | |
Mouse | |
Affinity chromatography | |
RUO | |
474543, 6819, 69083 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1A11 | |
Unconjugated | |
SULT1C2 | |
2610042J20Rik; E3 ubiquitin-protein ligase MGRN1; KIAA0544; mahoganoid; mahogunin ring finger 1; mahogunin ring finger 1, E3 ubiquitin protein ligase; mahogunin RING finger protein 1; mahogunin, ring finger 1; mahogunin, ring finger 1a; md; mgrn1; mgrn1a; mKIAA0544; nc; probable E3 ubiquitin-protein ligase MGRN1; RGD1311862; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1; RNF156; wu:fi38c03; zgc:55978 | |
Full length human recombinant protein of human SULT1C2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Mouse | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction