Learn More
Invitrogen™ MGRN1 Monoclonal Antibody (OTI3A6)

Description
MGRN1 Monoclonal Antibody for Flow, ICC/IF, Western Blot
Specifications
Specifications
| Antigen | MGRN1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI3A6 |
| Concentration | 0.67 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MGRN1 |
| Gene Accession No. | O60291 |
| Gene Alias | 2610042J20Rik; E3 ubiquitin-protein ligase MGRN1; KIAA0544; mahoganoid; mahogunin ring finger 1; mahogunin ring finger 1, E3 ubiquitin protein ligase; mahogunin RING finger protein 1; mahogunin, ring finger 1; mahogunin, ring finger 1a; md; mgrn1; mgrn1a; mKIAA0544; nc; probable E3 ubiquitin-protein ligase MGRN1; RGD1311862; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1; RNF156; wu:fi38c03; zgc:55978 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.