Learn More
MGRN1 Mouse anti-Human, Clone: OTI3C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502684

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.Specifications
MGRN1 | |
Monoclonal | |
Unconjugated | |
MGRN1 | |
RNF156 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23295 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60291 | |
MGRN1 | |
Full length human recombit protein of human MGRN1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.