Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGRN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530464 Supplier Invitrogen™ Supplier No. PA530464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (81%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGRN1
Applications Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol, 1% BSA and 0.01% thimerosal; pH 7
Gene MGRN1
Gene Accession No. O60291
Gene Alias 2610042J20Rik; E3 ubiquitin-protein ligase MGRN1; KIAA0544; mahoganoid; mahogunin ring finger 1; mahogunin ring finger 1, E3 ubiquitin protein ligase; mahogunin RING finger protein 1; mahogunin, ring finger 1; mahogunin, ring finger 1a; md; mgrn1; mgrn1a; mKIAA0544; nc; probable E3 ubiquitin-protein ligase MGRN1; RGD1311862; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1; RNF156; wu:fi38c03; zgc:55978
Gene Symbols MGRN1
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 67 and 294 of Human MGRN1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23295
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.