Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGST1 Monoclonal Antibody (14H2)
GREENER_CHOICE

Catalog No. PIMA534942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534942 Supplier Invitrogen™ Supplier No. MA534942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: SiHa cell, HepG2 cell, A549 cell, U937 cell, rat kidney tissue, human liver carcinoma tissue, human colon tissue, human placenta tissue, mouse liver tissue, MCF-7 cell.

The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGST1
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 14H2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene MGST1
Gene Accession No. P10620
Gene Alias 1500002K10Rik; glutathione S-transferase; glutathione S-transferase 12; Gst; GST12; MGC14525; MGST; mGST1; MGST-I; Microsomal glutathione S-transferase 1; microsomal glutathione S-transferase 1-like protein; microsomal glutathione-S-transferase; microsomal GST-1; Microsomal GST-I
Gene Symbols MGST1
Host Species Mouse
Immunogen Recombinant protein within Human MGST1 aa 1-144.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.