Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595753 Supplier Invitrogen™ Supplier No. PA595753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVDLTQVMDD EVFMAFASYA TIILSKMMLM STATAFYRLT RKVFANPEDC VAFGKGENAK KYLRTDDRVE RVRRAHLNDL ENIIPFLGIG LLYSLSGPDP; Positive Samples: Mouse lung; Cellular Location: Endoplasmic reticulum membrane, Microsome, Mitochondrion outer membrane, Multi-pass membrane protein, Peripheral membrane protein.

The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGST1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MGST1
Gene Accession No. P08011, P10620, Q91VS7
Gene Alias 1500002K10Rik; glutathione S-transferase; glutathione S-transferase 12; Gst; GST12; MGC14525; MGST; mGST1; MGST-I; Microsomal glutathione S-transferase 1; microsomal glutathione S-transferase 1-like protein; microsomal glutathione-S-transferase; microsomal GST-1; Microsomal GST-I
Gene Symbols MGST1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MGST1 (NP_0012474401).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171341, 4257, 56615
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.