Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGST2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542714 Supplier Thermo Scientific Supplier No. PA542714
Add to Cart
Add to Cart

MGST2 Polyclonal Antibody

MGST2 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen MGST2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MGST2
Gene Accession No. Q99735
Gene Alias FLJ27438; glutathione peroxidase MGST2; GST2; leukotriene C4 synthase MGST2; MGC14097; MGST2; MGST-II; Microsomal glutathione S-transferase 2; microsomal glutathione S-transferase II; microsomal GST-2; Microsomal GST-II; Unknown (protein for MGC:127901)
Gene Symbols MGST2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MGST2 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4258
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.