Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MGST2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552732
Description
Immunogen sequence: LKYKVTPPAV TGSPEFERVF RAQQNCVEFY PIF Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.
MGST2 (Microsomal glutathione S-transferase 2) is a 147 amino acid protein encoded by the human gene MGST2. MGST2 can catalyze the production of LTC4 (leukotriene C4) from LTA4 (leukotriene A4) and reduced glutathione. It can also catalyze the conjugation of 1-chloro-2,4-dinitrobenzene with reduced glutathione. MSGT2 is a multi-pass membrane protein found as a homodimer. MGST2 belongs to the MAPEG family and is expressed in liver, spleen, skeletal muscle, heart, adrenals, pancreas, prostate, testis, fetal liver, and fetal spleen. It has very low expression in lung, brain, placenta and bone marrow. Also, MGST2 is the only GST expressed in human umbilical vein endothelial cells (HUVECs).
Specifications
MGST2 | |
Polyclonal | |
Unconjugated | |
MGST2 | |
FLJ27438; glutathione peroxidase MGST2; GST2; leukotriene C4 synthase MGST2; MGC14097; MGST2; MGST-II; Microsomal glutathione S-transferase 2; microsomal glutathione S-transferase II; microsomal GST-2; Microsomal GST-II; Unknown (protein for MGC:127901) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99735 | |
MGST2 | |
Recombinant protein corresponding to Human MGST2. Recombinant protein control fragment (Product #RP-90627). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction