Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597969
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597969 Supplier Invitrogen™ Supplier No. PA597969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MGST3 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

The MAPEG family consists of six human proteins, several of which are involved the production of leukotrienes and prostaglandin E, important mediators of inflammation. This gene encodes an enzyme which catalyzes the conjugation of leukotriene A4 and reduced glutathione to produce leukotriene C4. This enzyme also demonstrates glutathione-dependent peroxidase activity towards lipid hydroperoxides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGST3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MGST3
Gene Accession No. O14880
Gene Alias 2010012L10Rik; 2010306B17Rik; 2700004G04Rik; AA516734; Glutathione peroxidase MGST3; GST-III; LTC4 synthase MGST3; MGST3; microsomal glutathione S-transferase; microsomal glutathione S-transferase 3; Microsomal glutathione S-transferase III; microsomal GST-3; Microsomal GST-III
Gene Symbols MGST3
Host Species Rabbit
Immunogen Recombinant Human Microsomal glutathione S-transferase 3 protein (30-70AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4259
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.