Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific MGV Vertical Mini-Gel Systems
SCI_ED

Catalog No. S05772 Shop All CBS Scientific Products
Change view
Click to view available options
Cooling Type:
Dual
Single
No. of Gels:
1
1 to 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cooling Type No. of Gels
S05772 Single 1
S05773 Dual 1 to 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05772 Supplier CBS Scientific Supplier No. MGV302

Includes

Single Unit:

  • Long-lasting silicone gasket to prevent buffer leakage during electrophoresis
  • Safety cover with attached power leads
  • 5 molded polypropylene spring clamps
  • Bussed together power leads
  • Platinum electrodes
  • 1 comb
  • 1 Gel Wrap Spacer Set
  • 1 Gel Wrap Gasket
  • 1 Gel Wrap Glass Plate Set
Double Unit:
  • Cooling chamber with coolant recirculation ports
  • Long-lasting silicone gaskets on both sides of unit to prevent buffer leakage during electrophoresis
  • Safety cover with attached power leads
  • 10 molded polypropylene spring clamps
  • Bussed together power leads
  • Platinum electrodes
  • 2 combs
  • 2 Gel Wrap Spacer Sets
  • 2 Gel Wrap Gaskets
  • 2 Gel Wrap Glass Plate Set

Available in single or dual units.

  • Accept most commercially available pre-cast gels
  • Two methods for gel casting:
    • Gel Wrap™: Precision-machined PVC spacers can be used with Gel Wrap for easy gel sealing
    • Gel Casting Stand: Two gels can be cast simultaneously
    • Multiple Gel Casting Chambers are available for casting either four or ten gels simultaneously
  • Voltage limit: 250VDC

Specifications
Construction:

  • Buffer chamber, safety cover: Acrylic
  • Electrodes: Platinum wire 0.012 in. diameter
  • Power cords: FR Urethane rated 7500VDC, 200mA, 65°C (149°F)
  • Combs: PTFE
  • Glass plates: Soda-lime glass
  • Spacers: PVC
  • Clamps: Polypropylene, stainless-steel

Single unit:

  • Small size permits rapid analysis of samples in short supply and reduces use of expensive reagents needed for gel formation and running buffer
  • Internal cooling chamber: No
Dual unit:
  • Provide the capability of running two gels simultaneously under identical buffer condition
  • Internal cooling chamber: Yes (for temperaturre control)

SDS PAGE, 2-D electrophoresis, agarose gel separations

Order Info

  • Single Units:
  • MGV-102: 7.75L x 5.51W x 5 in.H (19.7 x 14 x 12.7cm); Weight: 3 lb. (1.36kg)
  • MGV-302: 7.08L x 5.51W x 3.93 in.H (18 x 14 x 10cm); Weight: 3 lb. (1.36kg)
  • Dual Units:
  • MGV-202: 7.75L x 6.69W x 5 in.H (19.7 x 17 x 12.7cm); Weight: 5 lb. (2.26kg)
  • MGV-402: 7.08L x 5.9W x 3.93 in.H (18 x 15 x 10cm); Weight: 5 lb. (2.26kg)

Certifications

CE marked

Specifications

Certifications/Compliance CE
Cooling Type Single
Running Time 25 to 75 min.
Includes Electrophoresis unit, Silicone gaskets on front of unit, Platinum electrodes, safety cover with power Leads, 1 comb, 1 sets of Gel Wrap™ Glass plates, 1 sets of Gel Wrap™ spacers, 1 Gel Wrap™ gasket, 5
No. of Gels 1
Volume (English) Buffer 3.71 oz.
Volume (Metric) Buffer 110 mL
Width (English) 5.51 in.
Width (English) Gel 3.93 in.
Width (Metric) 14 cm
Width (Metric) Gel 10 cm
Height (English) 3.93 in.
Height (English) Gel 3.14 in.
Height (Metric) 10 cm
Depth (English) 7 in.
Depth (Metric) 18 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.