Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I Antibody (2G5), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065938AF488
Description
MHC Class I Monoclonal specifically detects MHC Class I in Human, Mouse, Rat, Porcine, Bovine, Chinese Hamster, Guinea Pig, Hamster, Sheep samples. It is validated for Flow Cytometry, Product Image.Specifications
MHC Class I | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HLA-E | |
Purified H-2Kb and H-2Db MHC-I molecules. | |
0.125 mL | |
Adaptive Immunity, Immunology, Major Histocompatibility Complexes | |
3133 | |
Human, Mouse, Rat, Pig, Bovine, Guinea Pig, Hamster, Sheep | |
Purified |
Flow Cytometry | |
2G5 | |
Flow Cytometry | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-65938 recognizes a monomorphic epitope on the mouse MHC class I antigen. The epitope recognized by clone 2G5 is conformation dependent and is reported to be phylogenetically conserved. Reactivity has been observed with some canine samples suggesting that this antibody may recognize a polymorphic epitope of canine MHC class 1. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction