Learn More
MHC Class I Antibody (A4), Alexa Fluor™ 647, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP143122AF647
Catalog No. NBP143122AH
Only null left
Add to Cart
Description
Description
MHC Class I Monoclonal specifically detects MHC Class I in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
MHC Class I
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
HLA-E
The immunogen for this antibody was HLA MHC Class I free chain without B2 microglobulin.
0.1 mL
Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology, Major Histocompatibility Complexes
3133
Store at 4C in the dark.
IgG1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
A4
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.