Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I Antibody (A4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP14312225UG
Description
MHC Class I Monoclonal specifically detects MHC Class I in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
MHC Class I | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunoprecipitation | |
A4 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
HLA-E | |
The immunogen for this antibody was HLA MHC Class I free chain without B2 microglobulin. | |
25 μg | |
Adaptive Immunity, Immunology, Major Histocompatibility Complexes | |
3133 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction