Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I Antibody (OX18), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1206405AF350
Description
MHC Class I Monoclonal specifically detects MHC Class I in Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
MHC Class I | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
HLA-E | |
Rat spleen glycoproteins | |
Primary | |
Recognizes a monomorphic determinant of rat MHC Class I (RT1A), expressed by all rat strains. However, quantitative measurements suggest that not all of the class I molecules are recognised. | |
Store at 4°C in the dark. |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OX18 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular) | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
0.1 mL | |
3133 | |
Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction