Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I (H-2Db) Monoclonal Antibody (28-14-8), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | MHC Class I (H-2Db) |
---|---|
Clone | 28-14-8 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M038T03Y08A
![]() |
Invitrogen™
M038T03Y08A |
100 μg |
Each for $375.00
|
|
|||||
M038T02Y08A
![]() |
Invitrogen™
M038T02Y08A |
25 μg |
Each for $194.00
|
|
|||||
Description
MHC Class I (H-2Db) Monoclonal antibody specifically detects MHC Class I (H-2Db) in Mouse samples. It is validated for Flow CytometrySpecifications
MHC Class I (H-2Db) | |
0.1 mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
P01895, P01896, P01897, P01899, P01900, P14426, P14427 | |
14964 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
28-14-8 | |
Flow Cytometry | |
NovaFluor Yellow 755 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein | |
IgG2a κ | |
Affinity chromatography | |
H2-D1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title