Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I (H-2Kb) Monoclonal Antibody (AF6-88.5.5.3), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$201.00 - $379.00
Specifications
Antigen | MHC Class I (H-2Kb) |
---|---|
Clone | AF6-88.5.5.3 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M052T03B08A
![]() |
Invitrogen™
M052T03B08A |
100 μg |
Each for $379.00
|
|
|||||
M052T02B08A
![]() |
Invitrogen™
M052T02B08A |
25 μg |
Each for $201.00
|
|
|||||
Description
MHC Class I (H-2Kb) Monoclonal antibody specifically detects MHC Class I (H-2Kb) in Mouse samples. It is validated for Flow CytometrySpecifications
MHC Class I (H-2Kb) | |
0.1 mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
P01901, P03991, P04223, P14428 | |
14972 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
AF6-88.5.5.3 | |
Flow Cytometry | |
NovaFluor Blue 660-120S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
IgG2a κ | |
Affinity chromatography | |
H2-K1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title