Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), eFluor™ 450, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

$349.00

Specifications

Antigen MHC Class I (H-2Kd/H-2Dd)
Clone 34-1-2S
Concentration 0.2 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-4190
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
48599882
100 μg
Each of 1 for $349.00
Only null left
Add to Cart
 
Description

Description

Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p. Applications Reported: This 34-1-2S antibody has been reported for use in flow cytometric analysis. Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications

Specifications

MHC Class I (H-2Kd/H-2Dd)
0.2 mg/mL
Flow Cytometry
eFluor 450
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
P01895, P01896, P01897, P01899, P01900, P01901, P03991, P04223, P14426, P14427, P14428
14964, 14972
Primary
Affinity chromatography
34-1-2S
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG2a κ
Mouse
H2-D1
class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, D-B alpha chain; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2K1; H2-K1; H2KD; histocompatibility 2, D region locus 1; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-D-q alpha-chain; MHC H2-K transplantation antigen; Q5k protein
H2-D1, H2-K1
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.