Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class I (H2-D1) Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144130
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144130 Supplier Invitrogen™ Supplier No. PA5144130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MHC Class I (H2-D1) Polyclonal Antibody for Western Blot, ELISA

Involved in the presentation of foreign antigens to the immune system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class I (H2-D1)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene H2-D1
Gene Accession No. P01895, P01896, P01897, P01899, P01900, P14426, P14427
Gene Alias D-D alpha chain; H-2 class I histocompatibility antigen; H-2D; H2-D1
Gene Symbols H2-D1
Host Species Rabbit
Immunogen Recombinant Mouse H-2 class I histocompatibility antigen, D-D alpha chain protein (25-311AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14964
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.