Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC class I Rabbit anti-Human, Clone: 10G4, Bioss
SDP

Catalog No. 501989705
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9705 Supplier Bioss Supplier No. BSM52827R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Like their mammalian counterparts, avian MHC Class I molecules, also known as B-F antigens, consist of a highly polymorphic alpha-chain noncovalently bound to the invariant beta2-microglobulin subunit. MHC Class I molecules are expressed on most nucleated cells where they present endogenously synthesized antigenic peptides to CD8+ T lymphocytes, which are usually cytotoxic T cells. The monoclonal antibody F21-2 also reacts with turkey MHC Class I.

Specifications

Antigen MHC class I
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 10G4
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene HLA-A
Gene Accession No. P04439
Gene Alias A29 protein; antigen presenting molecule; Aw-33; Aw-74; B12; BF; B-F; B-F histocompatibility F10 antigen; B-F major; B-F minor; BF1; BF12; BF2; BFa1; BFa2; B-F-beta-IV; B-FI; BFIV; B-FIV; BFIV21; B-F-S01; B-F-S02; B-F-S03; B-F-S04; B-F-S05; B-F-S06; B-F-S07; B-F-S09; B-F-S11; B-F-S12; B-F-S13; B-F-S14; BFw-03; BFw-05; BFz-01; B-LB2; BOLA class I histocompatibility antigen, alpha chain BL3-7-like; class I histocompatibility antigen, F10 alpha chain; F10 alpha chain; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leukocyte antigen class I-A; Major histocompatibility complex class I antigen BF2; major histocompatibility complex class I glycoprotein; major histocompatibility complex, class I, A; major histocompatibility complex, class I, G; MCH class I antigen; MHC BF1 class I; MHC BF2 class I; MHC class 1 antigen; MHC class I alpha chain 1; MHC class I alpha chain 2; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen alpha chain; MHC class I antigen B-F heavy chain; MHC class I antigen B-F major heavy chain; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class I glycoprotein; MHC class I molecule; MHC class II antigen; MHC classI antigen; OLA-I; OVAR; Ovar-I; ovar-MHCI-B564; ovar-MHCI-D5; ovar-MHCI-F12; truncated MHC class I antigen
Gene Symbols HLA-A
Host Species Rabbit
Immunogen Human MHC class I aa 1-200.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.