Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    MHC Class II Antibody (AP-MAB0874), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP106696MFV450
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF, NeutralizationSpecifications
| MHC Class II | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified | 
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF | |
| AP-MAB0874 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Activated C57BL/6 mouse spleen cells | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 3108 | |
| Store at 4C in the dark. | |
| IgG2 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            