Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Antibody (CVS20), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234848AF350
Description
MHC Class II Monoclonal specifically detects MHC Class II in Human, Bovine, Canine, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
MHC Class II | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
HLA-DMA | |
3132 cells | |
Primary | |
Human, Bovine, Canine, Equine | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
CVS20 | |
Flow Cytometry, Immunohistochemistry | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
Mouse | |
0.1 mL | |
3108 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction