Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Antibody (CVS20), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234848PCP
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Human, Bovine, Canine, Equine samples. It is validated for Flow CytometrySpecifications
MHC Class II | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Bovine, Canine, Equine, Rabbit | |
Purified |
Flow Cytometry, Immunohistochemistry | |
CVS20 | |
Flow Cytometry | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
3132 cells | |
0.1 mL | |
Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
3108 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction