Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Antibody (HIS19), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200462AF700
Description
MHC Class II Monoclonal specifically detects MHC Class II in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
MHC Class II | |
Monoclonal | |
Alexa Fluor 700 | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HIS19 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
HLA-DMA | |
The immunogen for this antibody is MHC Class II. | |
0.1 mL | |
Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators | |
3108 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction