Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC class II (I-A/I-E) Antibody (MK-D6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308856UV
Description
MHC class II (I-A/I-E) Monoclonal antibody specifically detects MHC class II (I-A/I-E) in Mouse, Human (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
MHC class II (I-A/I-E) | |
Monoclonal | |
DyLight 350 | |
E-alpha-f;H-2 class II histocompatibility antigen, E-K alpha chain;H-2 class II histocompatibility antigen, E-U alpha chain;H-2E;H2-Ea;H-2Ea;H2-Ea-ps;I;Ia;Ia3;Ia-3;I-E;I-Ealpha;MHC-H2-Ea | |
Spleen cells from B10.D2 mice | |
100 μg | |
Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
100504404 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry, Immunofluorescence | |
MK-D6 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse, Human (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction