Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II I-Ab Monoclonal Antibody (AF6-120.1), NovaFluor™ Red 685, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | MHC Class II I-Ab |
---|---|
Clone | AF6-120.1 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M046T03R02A
![]() |
Invitrogen™
M046T03R02A |
100 μg |
Each for $375.00
|
|
|||||
M046T02R02A
![]() |
Invitrogen™
M046T02R02A |
25 μg |
Each for $194.00
|
|
|||||
Description
MHC Class II I-Ab Monoclonal antibody specifically detects MHC Class II I-Ab in Mouse samples. It is validated for Flow CytometrySpecifications
MHC Class II I-Ab | |
0.1 mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
P01921, P06342, P06343, P06344, P06345, P06346, P14483 | |
14961 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
AF6-120.1 | |
Flow Cytometry | |
NovaFluor Red 685 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Abeta; AI845868; CELIAC1; DADB-249P12.2; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2Eb; H-2Eb; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; HLA-DQB; Ia2; Ia-2; Ia4; Ia-4; IAb; I-Abeta; IDDM1; major histocompatibility complex class II beta chain; MGC163794; MGC163796; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 | |
IgG2a κ | |
Affinity Chromatography | |
H2-Ab1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title