Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Mouse anti-Chicken, Pigeon, Caiman, R-PE, Clone: 2G11, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 835009
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
MHC Class II | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Flow Cytometry | |
2G11 | |
R-PE | |
Mouse | |
RUO | |
Chicken, Lizard, Pigeon | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction