Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II, Mouse anti-Human, Clone: TDR31.1, Abnova™

Catalog No. 89998964 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-964 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-964 Supplier Abnova Corporation Supplier No. MAB16190
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen MHC Class II
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone TDR31.1
Conjugate Unconjugated
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunohistochemistry (Frozen sections) Western Blot The optimal working dilution should be determined by the end user.
Formulation In 50 mM Na3PO4, 100 mM KCl, 150 mM NaCl, pH 7.5 (0.5 mg/mL Gentamicin sulfate).
Host Species Mouse
Immunogen Purified MHC Class II from JY cell line.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.