Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Polymorphic Antibody (CI.13.I), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064962
Description
MHC Class II Polymorphic Monoclonal specifically detects MHC Class II Polymorphic in Guinea Pig samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
MHC Class II Polymorphic | |
Monoclonal | |
Unconjugated | |
Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
Mouse | |
Tissue culture supernatant | |
RUO | |
Primary | |
Reacts with the MHC Class II epitope present on strain 13, but not strain 2 guinea pig cells. The antigen is also found in outbred Hartley guinea pigs. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CI.13.I | |
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
HLA-DRB1 | |
Guinea pig peritoneal T-cells | |
0.125 mL | |
Immunology, Major Histocompatibility Complexes | |
3123 | |
Guinea Pig | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction