Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II Rabbit anti-Human, Mouse, Rat, FITC, Polyclonal, FabGennix

Catalog No. 502207767
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7767 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7767 Supplier Fabgennix Inc Supplier No. MHCIIFITC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MHC Class II Polyclonal antibody specifically detects MHC Class II in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen MHC Class II
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate FITC
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias beta 1 domain; B-L; B-L beta chain; B-L beta minor; B-LB; BLB major; BLB1; BLB2; B-LB2; B-LB21; B-LB21 major; BLB3; Bl-Beta II protein; B-LBII; class II histocompatibility antigen, B-L beta chain; class II histocompatibility antigen, M alpha chain; D6S222E; DMA; FECA-DRB; GSP-BLB1; GSP-BLB2; HLA class II histocompatibility antigen, DM alpha chain; HLADM; HLA-DMA; major histocompatibility class II antigen B-L beta; major histocompatibility complex class II B; Major histocompatibility complex class II beta chain BLB1; Major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB2, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB3, (similar to HLA class II, D beta chain); major histocompatibility complex, class II, DM alpha; MHC 2; MHC Class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen B-F minor heavy chain; MHC class II antigen DMA; MHC class II antigen DRB; MHC Class II beta 1 and 2 domains; MHC class II beta 1 domain; MHC class II beta chain; MHC class II beta chain 1; MHC class II beta chain 2; MHC class II B-L beta chain; MHC II; MHCCII; MHCFECA-DRB; really interesting new gene 6 protein; RING6; S19; uncharacterized protein LOC101747454
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on mouse Major Histocomatibility Complex Class II antigen (MHC II).
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.