Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II RT1B Antibody (OX-6), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065541JF646
Description
MHC Class II RT1B Monoclonal antibody specifically detects MHC Class II RT1B in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating, CyTOFSpecifications
MHC Class II RT1B | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
OX-6 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, CyTOF-ready | |
DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC DPB1, RING6, RING7 | |
Rat thymocyte membrane glycoproteins | |
0.25 mL | |
Adaptive Immunity, Immunology | |
24738 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction