Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II RT1B Antibody (OX-6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB165541SS Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165541SS 0.05 mg
NB10065541 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165541SS Supplier R&D Systems Supplier No. NB10065541SS

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody has been used in 17 publications

MHC Class II RT1B Monoclonal specifically detects MHC Class II RT1B in Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MHC Class II RT1B
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OX-6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 31317505), CyTOF-ready
Formulation PBS with 0.02% Sodium Azide
Gene Alias anti-OX-6 RT1B, anti-RT1B OX-6, Clone OX6, DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC class II DQ, MHC DPB1, Non-polymorphic MHC Class II, RING6, RING7, RT1B
Gene Symbols RT1-B
Host Species Mouse
Immunogen Rat thymocyte membrane glycoproteins
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24738
Test Specificity NB100-65541 recognizes a monomorphic determinant of the rat I-A antigen present on B lymphocytes, dendritic cells, some macrophages and certain epithelial cells. This antibody cross reacts with certain mouse strains of MHC haplotypes k and s. This antibody does not react with C57BL6 or C57BL6/j mice because they are haplotype b. Analysis of recombinant mouse strains showed that the determinants mapped to the I-A region. The clone does not react with Rat BDIX strain. Please be aware of the possible size variance seen in western blot.
Target Species Mouse, Rat, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.