Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC I Free Chain Monoclonal Antibody (A4), eBioscience™, Invitrogen™

Catalog No. 14995837
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-995-837 2 mg
14-995-882 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-995-837 Supplier Invitrogen™ Supplier No. 14995837
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The A4 monoclonal antibody reacts with the class I human leukocyte antigen (HLA) complex in the absence of beta2 microglobulin. There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with beta2 microglobulin (beta2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. Applications Reported: This A4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This A4 antibody has been tested by flow cytometric analysis of 3-day PHA-activated normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with B2 microglobulin (B2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC I Free Chain
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone A4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias MHC Free
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.