Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC I Monomorphic Monoclonal Antibody (73-2)

MHC I Monomorphic Monoclonal Antibody (73-2)
Supplier: Thermo Scientific MA528478
Description
Specifications
| MHC I Monomorphic | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Rabbit | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 73-2 | |
| Unconjugated | |
| 3349.1; BOLA; BoLA-N; BoLA-NC; Bota; classical MHC class I antigen; D18.5; Eqca-1; EQMHCB2; EQMHCC1; HD15; hypothetical protein LOC509597; LA-A11; major histocompatibility complex; major histocompatibility complex, class I, A; MHC class 1; MHC class I antigen; MHC class I antigen 10.b; MHC class I antigen 2*02501; MHC class I antigen 2.5; MHC class I antigen 3.1; MHC class I antigen 3.5; MHC class I antigen 3.6; MHC class I antigen 9.5; MHC class I antigen 9.6; MHC class I antigen clone 2; MHC class I antigen heavy chain; MHC class I heavy chain; MHC class I heavy chain clone 302D.1m; MHCB1; MHCX1; non-classical MHC class I antigen; non-MHC class I antigen; uncharacterized protein LOC790811 | |
| Rabbit T cells. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction