Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC I Monomorphic Monoclonal Antibody (CVS22), FITC
GREENER_CHOICE

Catalog No. PIMA528475
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528475 Supplier Invitrogen™ Supplier No. MA528475
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Horse MHC Class I Monomorphic antibody, clone CVS22 recognizes monomorphic equine MHC Class I and was classified at the International Equine Leucocyte Antigen Workshop.

MHC I Monomorphic is part of major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC I Monomorphic
Applications Flow Cytometry
Classification Monoclonal
Clone CVS22
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene MHCX1
Gene Accession No. 0
Gene Alias 3349.1; BOLA; BoLA-N; BoLA-NC; Bota; classical MHC class I antigen; D18.5; Eqca-1; EQMHCB2; EQMHCC1; HD15; hypothetical protein LOC509597; LA-A11; major histocompatibility complex; major histocompatibility complex, class I, A; MHC class 1; MHC class I antigen; MHC class I antigen 10.b; MHC class I antigen 2*02501; MHC class I antigen 2.5; MHC class I antigen 3.1; MHC class I antigen 3.5; MHC class I antigen 3.6; MHC class I antigen 9.5; MHC class I antigen 9.6; MHC class I antigen clone 2; MHC class I antigen heavy chain; MHC class I heavy chain; MHC class I heavy chain clone 302D.1m; MHCB1; MHCX1; non-classical MHC class I antigen; non-MHC class I antigen; uncharacterized protein LOC790811
Gene Symbols EQMHCB2, EQMHCC1, MHCB1, MHCX1
Host Species Mouse
Immunogen Equine leucocytes.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100034209, 100034210, 100034211, 100056391
Target Species Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.