Learn More
Invitrogen™ MHC I Monomorphic Monoclonal Antibody (CVS22), PE

Description
Mouse anti Horse MHC Class I Monomorphic antibody, clone CVS22 recognizes monomorphic equine MHC Class I and was classified at the International Equine Leucocyte Antigen Workshop. Reconstitute by adding 1 mL distilled water.
Specifications
Specifications
| Antigen | MHC I Monomorphic |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CVS22 |
| Concentration | 1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 5% sucrose, 1% BSA and 0.09% sodium azide |
| Gene | MHCB1 |
| Gene Accession No. | 0 |
| Gene Alias | 3349.1; BOLA; BoLA-N; BoLA-NC; Bota; classical MHC class I antigen; D18.5; Eqca-1; EQMHCB2; EQMHCC1; HD15; hypothetical protein LOC509597; LA-A11; major histocompatibility complex; major histocompatibility complex, class I, A; MHC class 1; MHC class I antigen; MHC class I antigen 10.b; MHC class I antigen 2*02501; MHC class I antigen 2.5; MHC class I antigen 3.1; MHC class I antigen 3.5; MHC class I antigen 3.6; MHC class I antigen 9.5; MHC class I antigen 9.6; MHC class I antigen clone 2; MHC class I antigen heavy chain; MHC class I heavy chain; MHC class I heavy chain clone 302D.1m; MHCB1; MHCX1; non-classical MHC class I antigen; non-MHC class I antigen; uncharacterized protein LOC790811 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.