Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC I polymorphic Monoclonal Antibody (ER-MP42), Invitrogen™

Catalog No. MA181746
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA181746 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA181746 Supplier Thermo Scientific Supplier No. MA181746

Rat Monoclonal Antibody

Clone ER-MP42 specifically recognizes mouse strains with the haplotypes H-2Kv, H-2Dd and H-2k, q, s. Mouse strains with the haplotypes H-2p, r, w7, w22 show weak reactivity with this antibody. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

The major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. In mice, this complex is referred to as the histocompatibility 2 (H-2) region. The polymorphic epitope is present on murine MHC class I molecules, which are expressed at varying levels on the majority of nucleated cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC I polymorphic
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-MP42
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene H2-K1
Gene Accession No. P01895, P01896, P01897, P01899, P01900, P01901, P03991, P04223, P14426, P14427, P14428
Gene Alias blastocyst MHC; class I major histocompatibility antigen H-2Kd; H2 blastocyst; H-2 class I histocompatibility antigen, D-B alpha chain; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H2-B1; H2-Bl; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2-D1; H2-DC1-beta transplantation antigen protein; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, blastocyst; histocompatibility 2, D region locus 1; histocompatibility 2, K1, K region; K-f; MHC Class 1; MHC class I antigen; MHC class I antigen splice variant Bl.1; MHC class I H2 antigen; MHC class I H-2Dp; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-D-q alpha-chain; MHC H2-K transplantation antigen; Q5k protein
Gene Symbols H2-Bl, H2-D1, H2-K1
Host Species Rat
Immunogen Balb/c macrophage precursor cell hybrids.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14963, 14964, 14972
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.