Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC II (HLA-DP and DR) Monoclonal Antibody (ABM288), Abbomax

Catalog No. 502203770
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3770 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3770 Supplier Abbomax Inc Supplier No. 5006144
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Tonsil/lymph node Cellular location: Membrane.

MHC II (HLA-DP and DR) Monoclonal antibody specifically detects MHC II (HLA-DP and DR) in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed)

Specifications

Antigen MHC II (HLA-DP and DR)
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone ABM288
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene HLA-DPB1
Gene Accession No. P01911, P04440
Gene Alias beta1 domain MHC class II HLA DPB; class II HLA beta chain; Clone P2-beta-3; DPB1; DR-16; DRB1; DRw10; DW2.2/DR2.2; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; lymphocyte antigen DRB1; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DR beta 1; MHC Class 2; MHC class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB
Host Species Mouse
Immunogen Activated human peripheral blood mononuclear cells
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3115, 3123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.